跳转至内容
Merck
  • DNA cross-linking and sequence selectivity of aziridinylbenzoquinones: a unique reaction at 5'-GC-3' sequences with 2,5-diaziridinyl-1,4-benzoquinone upon reduction.
DNA cross-linking and sequence selectivity of aziridinylbenzoquinones: a unique reaction at 5'-GC-3' sequences with 2,5-diaziridinyl-1,4-benzoquinone upon reduction.
Biochemistry (1991-12-17)
J A Hartley, M Berardini, M Ponti, N W Gibson, A S Thompson, D E Thurston, B M Hoey, J Butler
摘要
Several bifunctional alkylating agents of the aziridinylbenzoquinone class have been evaluated as potential antitumor agents. 3,6-Bis[(2-hydroxyethyl)amino]-2,5- diaziridinyl-1,4-benzoquinone (BZQ), 2,5-diaziridinyl-1,4-benzoquinone (DZQ), 3,6-bis(carboxyamino)-2,5-diaziridinyl- 1,4-benzoquinone (AZQ), and six analogues of AZQ have been studied for their ability to induce DNA interstrand cross-linking, as measured by an agarose gel technique, and to determine whether they react with DNA in a sequence-selective manner, as determined by a modified DNA sequencing technique. At an equimolar concentration (10 microM), only DZQ and BZQ showed any detectable cross-linking at pH 7 without reduction. Cross-linking was enhanced in both cases at low pH (4). Reduction by ascorbic acid at both pH's increased the cross-linking, which was particularly striking in the case of DZQ. In contrast, AZQ and its analogues only produced a significant level of cross-linking under both low-pH and reducing conditions, the extent of cross-linking decreasing as the size of the alkyl end group increased. The compounds reacted with all guanine-N7 positions in DNA with a sequence selectivity similar to other chemotherapeutic alkylating agents, such as the nitrogen mustards, although some small differences were observed with BZQ. Nonreduced DZQ showed a qualitatively similar pattern of reactivity to the other compounds, but on reduction (at pH 4 or 7) was found to react almost exclusively with 5'-GC-3' sequences, and in particular, at 5'-TGC-3' sites. A model to explain this unique reaction is proposed.