主要文件
CLS6624
Corning® Step-R Repeating Pipettor Syringe Tips
volume (5.0 mL), non-sterile
别名:
pipet tips
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121600
NACRES:
NB.02
无菌性
non-sterile
包装
pack of 100 ea
制造商/商品名称
Corning 6624
体积
(5.0 mL)
正在寻找类似产品? 访问 产品对比指南
一般描述
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
法律信息
Corning is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)