YY1601209
1 of 10
Millipore
XX6700010
Cylinder O-rings
YT30142HW
有害废物过滤系统,142 mm
Milli-Q
CF0F01205
Kit: Milli-Q® Organic-Free (RO feedwater)
Supelco
1.01517
Silver sulfate solution 10 g/l
KGEPS015FF3
Opticap XL 150 Millipore Express SHF 无菌 0.2 µm 3/4 in.TC/TC
Sigma-Aldrich
431648
硫化钠 九水合物
XX6700059
O-ring, fluoroelastomer A
ICS023
硫化物IC标准品
419273
羧甲基纤维素钠
HAWP04700
MF-Millipore™ 滤膜,孔径0.45 µm
for use with PL-1 Cartridge Housings
Filter Housings