主要文件
XX6320022
Millipore
Battery Charger
for use with Millipore Single/Dual Chamber Incubator, AC/DC input 230 V, 50 Hz
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104424
质量水平
应用
bioburden testing
cosmetics
environmental
food and beverages
相容性
for use with Millipore Single/Dual Chamber Incubator
一般描述
The Battery Charger for Single/Dual chamber incubator is designed to charge the rechargeable nickel- cadmium battery installed in the incubator.