SPPS00006-M
1 of 10
Millipore
17403X1
Cartridge Housing Silicone Vent/Drain Valve Seal Kit
D00271V01
Clarigard 筒式过滤器 10 in.0.2 µm 7 号 氟橡胶
KGEPA10TT1
Opticap XL 10 Millipore Express SHF 可高压蒸汽灭菌 0.2 µm 1-1/2 in.TC/TC
MXPESP5CN
阀门出口帽+针头
MXPESPTCL
MicropreSure® 卫生级取样阀
Milli-Q
ZWDK5L100
垫圈分配套件
CWSS71S03
Milligard® 0.5/0.2 µm标称,小柱
Sigma-Aldrich
202614
硫酸铝
YY2229362-M
O-ring Support Screen fluoroelastomer for 293 mm Filter Holder
CKT1MAG-97K
MILLIPLEX® Canine Kidney Toxicity Expanded Magnetic Bead Panel 1, CKT1MAG-97K
silicone
100
for use with OPTISEAL® Sanitary Cartridge Housings
Filter Housings
Filtration
Filtration for Bioprocessing
Sterile Filtration and Bioburden Control