MSP000993
1 of 10
Millipore
PICM01250
Millicell® Standing细胞培养池
MSP000950
Durapore® 0.22 µm,小柱
MSP000953
MSP000994
FLUOROPORE FSLW 35MM x 12M ROLL QTY/1
Sigma-Aldrich
NS220
Neurite Outgrowth Assay Kit (3 µm)
GVWP2932A
Durapore®筛网过滤器
GVWP04700
Durapore®膜过滤装置, 0.22 µm
BGCM00010
PTFE Membrane Filter, 0.4 μm Pore Size
MSP000995
Fluoropore FSLW 34mm X 12m
HAWP04700
MF-Millipore™ 滤膜,孔径0.45 µm
400