1.51432
1 of 10
Supelco
1.50470
Purospher® STAR RP-18 封端 (3 µm)
1.51486
manu-CART® NT卡套,用于LiChroCART® 2,3,4,4.6 mm i.d.HPLC 柱
1.51456
Purospher® STAR RP-18封尾 (5 µm) Hibar® RT 250-4.6
50139-U
LiChrospher® RP-18 HPLC 色谱柱
1.50359
Purospher® STAR RP-18封尾 (5µm) LiChroCART® 250-4.6
1.50358
Purospher® STAR RP-18 封端(5µm) LiChroCART® 150-4.6
1.51455
Purospher® STAR RP-18 封端 (5µm) Hibar® 150-4.6
1.06346
一水磷酸二氢钠
1.50250
Purospher® STAR RP-18 封端(5 µm)LiChroCART® 4-4
1.50031
Purospher® STAR RP-8封端(5µm) LiChroCART® 150-4.6
LiChroCART® 150-4, HPLC cartridge
100