主要文件
Z677949
Finntip® Flex pipette tips
Finntip Flex 300, non-sterile
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121600
NACRES:
NB.01
物料
colorless tip
orange
无菌性
non-sterile
特点
barrier: no
包装
pack of 1000 ea (Bag)
制造商/商品名称
Finn 94060520
最大体积
300 μL
体积范围
5-300 μL
正在寻找类似产品? 访问 产品对比指南
一般描述
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
其他说明
Browse all pipette tips
法律信息
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
历史批次信息供参考:
分析证书(COA)