主要文件
Z507954
Recovery flask
capacity 10 mL
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121804
NACRES:
NB.43
物料
glass
round bottom flask
single-neck
无菌性
non-sterile
特点
closure type Holed cap
包装
pack of 1
制造商/商品名称
Ace Glass 9470-43
容量
10 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
<ST/>14/10 threaded joint, and modified sides for easy cleaning or removal of material. Supplied with holed cap, PTFE-faced silicone septum, and FETFE® O-ring.
法律信息
FETFE is a registered trademark of Ace Glass, Inc.
历史批次信息供参考:
分析证书(COA)