主要文件
Z405558
J. Young high-vacuum PTFE valve, straight through
stopcock size 4 mm, side arm O.D. 7 mm
登录查看公司和协议定价
About This Item
UNSPSC代码:
40141600
NACRES:
NB.79
制造商/商品名称
J. Young
侧臂外径
7 mm
活塞尺寸
4 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
The J. Young high-vacuum PTFE valve helps in controlling the atmosphere when using air-sensitive materials.
Features:
  • PTFE piston with unique PTFE-sheathed elastomer O-ring seals
  • Grease-free glass-to-PTFE seals
  • High-vacuum to 10-6 torr
历史批次信息供参考:
分析证书(COA)