主要文件
Z206725
Sigma-Aldrich
Scienceware® polyethylene dipper
handle L 6 ft
登录查看公司和协议定价
About This Item
UNSPSC代码:
41100000
NACRES:
NB.51
描述
one-piece
制造商/商品名称
Bel-Art
把手长度
6 ft
正在寻找类似产品? 访问 产品对比指南
一般描述
Corrosion-free, durable HDPE construction will not contaminate solutions. 11/8 in. diam. handle is welded at a 45° angle to a rigid 4 in. diam., 16 oz bowl allowing for sampling in hard-to-reach areas.
法律信息
Scienceware is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)