Merck
Z202312
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 29/15
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122413
NACRES:
NB.35
材料
stainless steel
接合處
joint: SJ29/15
正在寻找类似产品? 访问 产品对比指南
一般說明
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.