Z173088
1 of 10
Sigma-Aldrich
Z166898
Aldrich® 气敏化学玻璃实验室套件
Z173096
Solvent storage flask with glass stopcock on side-arm
Z122548
Aldrich® student glass lab kit
Z220302
Reaction flask with glass stopcock on side-arm
Z173061
Z515981
Schlenk reaction and storage tube
SYNF484250
Synthware™ solvent transfer/storage flask
Z516082
Z515868
Aldrich® solvent storage flask
Z515884
borosilicate glass round bottom two-neck
S
female joint: ST/NS14/20
non-sterile
closure type female Joint: ST/NS
pack of 1
100 mL
正在寻找类似产品? 访问 产品对比指南
实验室容器与储存
实验室烧瓶
样品处理
玻璃耗材
批号
在文件库中查找您最近购买产品的文档。
访问文档库
1 of 4
Z515701
Schlenk Kjeldahl reaction and storage flask
Z173231
Reaction and storage flask, Kjeldahl, with glass stopcock on side-arm
Z516015
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.