Z112143
1 of 10
Sigma-Aldrich
203033
碘化铯
Z123633
圆形晶体窗片
W266523
L-薄荷醇
383805
1,4,8,11,15,18,22,25-八丁氧基-29H,31H-酞菁
W266590
M2772
薄荷醇
Supelco
63660
(−)-薄荷醇
224464
D-薄荷醇
202134
805874
n-碘化丁胺
Irtran-2
pkg of (contains 1 window)
IR spectroscopy: suitable
32 mm × 2 mm
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.