主要文件
TEM-CF313-50
Sigma-Aldrich
C-flat Holey Carbon Grids (CF-1.2/1.3) for TEM
Copper Grids, size 300 mesh, pack of 50
登录查看公司和协议定价
About This Item
UNSPSC代码:
41111761
NACRES:
MA.24
包装
pack of 50
尺寸
300 mesh
正在寻找类似产品? 访问 产品对比指南
一般描述
C-flat Holey Carbon Grids for TEM, Copper Grids, 300 Mesh, 1.2 micron hole, 1.3 micron spacing, 50/pk. C-flat is a holey carbon support film, manufactured without plastics, resists or other soft materials. As a result, the carbon films are flat, uniform
历史批次信息供参考:
分析证书(COA)