主要文件
SYNA351410
Synthware distillation adapter
joint: ST/NS 14/10
别名:
Synthware A351410, adapter, sidearm adapter
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
NACRES:
NB.51
接合处
joint: ST/NS14/10
包装
pack of 1 ea
制造商/商品名称
A351410
相关类别
一般描述
Synthware distillation side arm adapter with 14/10 joint. Supplied with one holed compression cap and one ViltonTM O-ring.
法律信息
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)