主要文件
DWK73800-15415
KIMBLE® Solid Cap
with rubber liner, phenolic, cap for 15-415
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
NACRES:
NB.11
物料
PTFE/rubber liner
无菌性
non-sterile
特点
class A
包装
case of 1000 ea
制造商/商品名称
Kimble® 73800-15415
配件
cap for 15-415
正在寻找类似产品? 访问 产品对比指南
一般描述
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
Identified in ASTM Specification E982, Class A requirement
特点和优势
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirement
法律信息
KIMBLE is a registered trademark of DWK Life Sciences
历史批次信息供参考:
分析证书(COA)