Merck
DWK14395-100
KIMBLE® KIMAX® Media bottle
with cap, round transparent borosilicate glass bottle, bottle capacity (100 mL), non-sterile
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
NACRES:
NB.43
物料
blue polypropylene cap
round transparent borosilicate glass bottle
无菌性
non-sterile
特点
with cap
包装
case of 10 ea
制造商/商品名称
Kimble® 14395-100
瓶容量
(100 mL)
配件
GL 45 neck
GL 45
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
GL 45 Media Bottles ideal for general laboratory use including mixing, storing or transporting culture media, chemicals or solvents.
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Replaceable clear (natural) drip-free polypropylene pour ring is included on each bottle
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
特点和优势
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
法律信息
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
历史批次信息供参考:
分析证书(COA)