主要文件
BR452210
BRAND®一次性接种环,带针头
volume 10 μL, blue
别名:
接种环
登录查看公司和协议定价
About This Item
UNSPSC代码:
41161502
NACRES:
NB.22
物料
blue
polystyrene
无菌性
sterile; γ-irradiated
包装
pack of 1000 ea
pkg of 1000 ea (50 bags of 20)
制造商/商品名称
BRAND 452210
体积
10 μL
正在寻找类似产品? 访问 产品对比指南
一般描述
For inoculation of nutrient media. Available either with loop at one end and needle at the other or loops at both ends. High flexibility permits gentle streaking without damaging the nutrient surface. γ-ray sterilized.
特点和优势
  • Sterile (SAL 10-6)
  • High flexibility for agar-saving smears
  • Usable at both ends
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)