主要文件
BR13908
BRAND®紧凑型SILBERBRAND滴定管,带PTFE旋塞
volume 50 mL
别名:
SILBERBRAND, 滴定管, 紧凑型滴定管
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121808
NACRES:
NB.43
物料
glass
制造商/商品名称
BRAND 13908
体积测量范围
50 mL, accuracy: ±0.10 mL
体积
50 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
The New Generation of Burettes from BRAND. With PTFE stopcock, fast to dismantle and easy to clean. Simple repair - all individual components replaceable. Boro 3.3, removable PTFE stopcock. Error limits according to class B, DIN EN ISO 385. Calibrated to deliver (TD, Ex).
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)