CLS3801
Corning® Costar® 12 mm , 10 μm pore size , 1.12 cm2 , cell growth area plates
0.4 μm pore size clear polystyrene plate, polyester membrane, flat bottom wells clear sterile, pack of 6, case of 24
Synonym(s):
cell culture inserts
Select a Size
Select a Size
About This Item
Recommended Products
Product Name
Corning® Costar® Snapwell cell culture inserts, 12 mm Snapwell Insert with 0.4 μm pore, pore density 4 x 106 pores/cm2, polyester membrane, TC-treated, sterile, 24/cs
material
clear polystyrene plate
flat bottom wells clear
polyester membrane
sterility
sterile
feature
lid
plate format: 6 well
packaging
case of 24
pack of 6
manufacturer/tradename
Corning 3801
membrane diam.
12 mm
membrane thickness
10 μm
surface area
1.12 cm2 , cell growth area
pore size
0.4 μm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- A modified Transwell permeable support containing a 12 mm diameter 0.4μm pore polyester membrane supported by a detachable ring
- Once cells are grown to confluence on the Snapwell insert, the ring can be placed in a vertical or horizontal diffusion chamber.
- Sterilized by gamma radiation
- Packaged in 6 well plates
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service