Skip to Content
Merck
All Photos(1)

Key Documents

57278

Supelco

Stainless Steel Weights For Visiprep Large Volume Sampler

weight fitting for 1/8 in. O.D. tubing, pk of 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$448.92

$448.92

List Price$522.00Save 14%

Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB04901

To order products, please contact your local dealer.

Select a Size

Change View
100 μG
$448.92

About This Item

UNSPSC Code:
12000000

$448.92

List Price$522.00Save 14%

Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB04901

To order products, please contact your local dealer.

material

stainless steel weights

packaging

pk of 4

technique(s)

solid phase extraction (SPE): suitable

fitting

weight fitting for 1/8 in. O.D. tubing

application(s)

food and beverages

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB5200988SAB5200888SAB5200904
conjugate

unconjugated

conjugate

Allophycocyanin conjugate

conjugate

unconjugated

conjugate

phycoerythrin (R-PE) conjugate

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

human, mouse, rat

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

clone

S327A-38, monoclonal

clone

S327A-38, monoclonal

clone

S327-95, monoclonal

clone

S327-95, monoclonal

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

General description

Stainless steel weights for Visiprep large volume are components that are fitted at one end of visiprep large volume samplers, which also consists of 1/8-inch polytetrafluoroethylene (PTFE) tubing with a screw fitting at the other end.

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service