Skip to Content
Merck
All Photos(3)

Key Documents

SAB2702172

Sigma-Aldrich

Monoclonal Anti-XPC antibody produced in mouse

Synonym(s):

RAD4, XP3, XPCC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ICC
IF
IHC (p)
WB
clone:
3.26
monoclonal
species reactivity:
human, mouse
citations:
technique(s):
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 500-3000

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3.26
monoclonal

form

liquid

mol wt

120-125 kDa

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 500-3000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XPC(7508)

Immunogen

Recombinant XPC purified from E. coli

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: Assay-dependent dilution, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Human XPC (Xeroderma pigmentosum group C) is a member of a family of proteins that has been shown to be involved in the repair of DNA via the nucleotide excision repair (NER) pathway. Specifically, XPC is believed to be a part of a heteromeric protein complex that is involved in the recognition of the DNA lesions during global genomic repair but not transcription-coupled repair.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service