Merck
23814
Supelco
Herramienta capilar Cleaving
Retractable Blade Version
About This Item
Código UNSPSC:
41115700
Materiales
sapphire cutting blade
envase
pkg of 1 ea
Descripción general
Capillary Cleaving Tool increases the column performance by making the properly sized, squarely-cut ferrule to seal the capillary column end.
This handy tool makes scalpel-like cuts to polyimide-coated fused silica, leaving no jagged edges to create problems. The industrial sapphire cutting edges remains sharp indefinitely.
Otras notas
The spring-loaded retractable blade reduces the chance of breakage if the tool is dropped.
Información legal
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)