Documentos clave
BAF369310000
Bel-Art® Earth Friendly 1001 Stick
package of 100
Seleccione un Tamaño
100 EA
82,70 €
82,70 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
100 EA
82,70 €
About This Item
Código UNSPSC:
41122403
NACRES:
NB.35
82,70 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Materiales
( PLA (polylactic acid) resin )
envase
package of 100
fabricante / nombre comercial
Bel-Art F36931-0000
L × An × Al
5 3/4 in. × 3/4 in. × 1/16 in.
Descripción general
Lessen environmental impact with these PLA (polylactic acid) resin sampling sticks. The production of 100% plant-based PLA contributes less greenhouse gasses and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Whether you are mixing samples, handling capsules, scraping containers or positioning solid samples for examination, 1001 Sticks are ready to assist! Use a separate stick for each material sampled to avoid cross contamination.
Características y beneficios
Green alternative to petroleum-based sampling tools; helps meet sustainability goals
Comparable in strength and durability to polystyrene, polypropylene and other petroleum-based plastic resin sampling tools
Disposal options include industrial composting for biodegradability, incineration, recycling and landfill
Información legal
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)