Skip to Content
Merck
All Photos(2)

Documents

90568

Supelco

Fenofibric acid

analytical standard

Synonym(s):

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, Fenofibric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClO4
CAS Number:
Molecular Weight:
318.75
Beilstein:
2058973
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

O=C(C1=CC=C(OC(C)(C)C(O)=O)C=C1)C2=CC=C(Cl)C=C2

InChI

1S/C17H15ClO4/c1-17(2,16(20)21)22-14-9-5-12(6-10-14)15(19)11-3-7-13(18)8-4-11/h3-10H,1-2H3,(H,20,21)

InChI key

MQOBSOSZFYZQOK-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

Application

Fenofibric acid may be used as a reference standard in the determination of fenofibric acid in samples of human plasma using high performance liquid chromatography (HPLC) and liquid chromatography coupled with tandem mass spectrometry (LC-MS-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simultaneous determination of rosuvastatin and fenofibric acid in human plasma by LC?MS/MS with electrospray ionization: assay development, validation and application to a clinical study.
Trivedi K R, et al.
Journal of Pharmaceutical and Biomedical Analysis, 39(3-4), 661-669 (2005)
Determination of bezafibrate, ciprofibrate and fenofibric acid in human plasma by high-performance liquid chromatography.
Masnatta D L, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 687(2), 437-442 (1996)
Martial Caillaud et al.
Cancers, 13(1) (2021-01-02)
Paclitaxel-induced peripheral neuropathy (PIPN) is a major adverse effect of this chemotherapeutic agent that is used in the treatment of a number of solid malignancies. PIPN leads notably to burning pain, cold and mechanical allodynia. PIPN is thought to be
Roger K Verbeeck et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 18(1), 61-67 (2015-04-17)
The therapeutic equivalence of multiple registered fenofibrate formulations, several of which are suprabioavailable and therefore marketed at lower dosage strengths than their reference products, is based on the results of bioequivalence studies. Most of these formulations show a higher bioavailability
Yunxia Lu et al.
Biochemical and biophysical research communications, 458(1), 1-7 (2015-01-17)
Fenofibrate (FF) is widely used to lower blood lipids in clinical practice, but whether its protective effect on endothelium-dependent vasodilatation (EDV) in thoracic aorta is related with endoplasmic reticulum (ER) stress remains unknown. In this study, female Sprauge Dawley rats

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service