Z710229
Nichipet glass tip for Eco pipette
volume range 20-200 μL
About This Item
Productos recomendados
material
colorless
glass
sterility
non-sterile
feature
barrier: no
packaging
pack of 200 ea (Racked)
manufacturer/tradename
Nichiryo 00-GLT-S
availability
not available in (Not available in France)
maximum volume
200 μL
volume range
20-200 μL
Categorías relacionadas
For use with
Referencia del producto