Documentos
Z708275
Slide mailer, 2 place
natural
About This Item
UNSPSC Code:
41123302
NACRES:
MA.01
material
polypropylene
feature
Holds 2 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15983B
size
93x68x50 mm
color
natural
General description
These 2-place slide protectors hold standard slides (3 × 1 in. or 25 × 75 mm) securely. With large, easy to secure snaps our mailers provide safe storage for valuable slides. The bright neon colors bring some fun to the lab and come in handy for color-coding