Documentos clave
Z369071
Finnpipette® Stepper pipette tips
maximum volume 1.25 mL
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
colorless
sterility
non-sterile
feature
barrier: no
packaging
pack of 100 ea
manufacturer/tradename
Finn 9404180
maximum volume
1.25 mL
General description
Delivers 25-125 μl
Seven sizes, each available in bulk pack or sterile single-wrapped.
Packaging
bulk pack
Other Notes
Browse all pipette tips
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries