Skip to Content
Merck
All Photos(3)

Key Documents

698172

Sigma-Aldrich

Iron(II) trifluoromethanesulfonate

greener alternative

≥85%

Synonym(s):

Iron(II) triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2F6FeO6S2
CAS Number:
Molecular Weight:
353.98
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥85%

form

powder

reaction suitability

core: iron
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>300 °C

greener alternative category

SMILES string

FC(F)(F)S(=O)(=O)O[Fe]OS(=O)(=O)C(F)(F)F

InChI

1S/2CHF3O3S.Fe/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2

InChI key

PGJLOGNVZGRMGX-UHFFFAOYSA-L

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service