Skip to Content
Merck
All Photos(1)

Documents

BSC1

85011422, African green monkey kidney, Epithelial | Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BSC1, 85011422, from African green monkey kidney, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

African green monkey kidney

growth mode

Adherent

karyotype

60

morphology

Epithelial
Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Monkey African green kidney epithelial

Cell Line Description

Continuous cell line which when infected with SV40 virus develops characteristic cytopathic effect.

Culture Medium

EMEM (EBSS) (M2279) + 2 mM L-Glutamine (G7513) + 1% NEAA (M7145) + 10% FBS / FCS (F2442).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service