Skip to Content
Merck
All Photos(1)

Documents

m17.ASC

11112201, mouse adipose tissue, Fibroblast-like

Synonym(s):

m17 ASC, m17ASC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

m17.ASC, 11112201

biological source

mouse adipose tissue

growth mode

Adherent

karyotype

Normal diploid

morphology

Fibroblast-like

receptors

Sca-1, c-kit/CD117, nestin, nucleostemin, CD44 and CD106

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Adipose cell line, mouse subcutaneous, mesenchymal, multipotent

Cell Line Description

m17.ASC is a spontaneously immortalized multipotent mesenchymal cell line capable of acquiring adipocyte, chondrocyte, osteocyte and cardiomyocyte phenotypes when cultured with appropriate differentiation factors. m17.ASC provides a valuable tool for in vitro biomimetic studies; assessing growth technologies such as 3-D structures, biochemical cues (selected growth factors and hormones) and physical forces (electrical, mechanical and topographical). It can be used to screen compounds that induce or inhibit proliferation and/or differentiation in tissue specific phenotypes.

DNA Profile

Not specified

Culture Medium

51800C (51800C) + L-Glutamine (G7513) + FBS / FCS (F2442).

Subculture Routine

Split sub-confluent cultures (90%) approximately 1:6 with media changes or subculture every 2-3 days seeding at 0.5-4 x 104 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service