Merck
Z665622
Ohaus® Explorer® Pro Waage
EP114C, AC/DC input 230 V AC
Synonym(e):
laboratory balance, weighing balance
About This Item
UNSPSC-Code:
41111502
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Leistungsmerkmale
CE compliant
CSA approved
Hersteller/Markenname
EP114C
Verfügbarkeit
available only in EU
Messbereich Wiegekapazität
110 g, resolution: 1 mg
Plattformdurchm.
90 mm
Verwandte Kategorien
Allgemeine Beschreibung
The Explorer Pro Series of standard level balances was designed for simplicity and includes the industry′s easiest user interface for easy setup and operation; no training is required.
  • Simple 2 key menu operation and navigation
  • Text prompts for fast setup
  • 7 Application modes plus below balance weighing
  • Brilliant backlit high resolution dot matrix display
  • RS232 port for easy communication with computer or printer
  • GLP compliant with user selectable outputs
  • 5 Language operational display text
  • Protective in-use cover and security bracket included
  • Industry leading warranty and support
Rechtliche Hinweise
Explorer is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)