SYNA232419
Synthware™ anti-splash adapter with fritted disc
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 19/22
Synonym(e):
Synthware A232419, rotary evaporator bump trap
Empfohlene Produkte
Verbindung
bottom joint: ST/NS19/22
joint: ST/NS top24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A232419
Kapazität
100 mL
Verwandte Kategorien
Allgemeine Beschreibung
With extra coarse fritted disc sealed above lower inner joint. Frit reduces foam from being carryover.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.