Merck
Z380245
Jars Nalgene® style 2118
capacity 250 mL
Synonym(e):
laboratory jars, nalgene jars
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
polypropylene bottle
polypropylene cap
round translucent bottle
Verbindung
Joints threaded neck (70)
Beschreibung
Autoclavable with closure completely unscrewed
Leistungsmerkmale
with cap
Verpackung
pkg of 6
Hersteller/Markenname
Nalgene® 2118-0008
Kapazität
250 mL
Verwandte Kategorien
Allgemeine Beschreibung
Translucent with excellent chemical resistance.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries