Merck
TMO362008-9025
Nalgene® diagnostic bottle without cap
white high-density polyethylene, volume 8 mL, case of 2000 ea
Synonym(e):
PPCO bottles, small volume bottle
Größe auswählen
2000 EACH
1.080,00 €
1.080,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
2000 EACH
1.080,00 €
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
1.080,00 €
Check Cart for Availability
Materialien
white high-density polyethylene
Verpackung
case of 2000 ea
Hersteller/Markenname
Nalgene 362008-9025
Kapazität
8 mL
Volumen
8 mL
Allgemeine Beschreibung
Safely store and ship light-sensitive diagnostic samples and valuable reagents in small volumes with Thermo Scientific Nalgene Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque amber high-density polyethylene bottles are for use with light sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)