Merck
TMO3405-42
Nalgene®PC Biotainer® Bottles and Carboys
clear polycarbonate, 5000 mL capacity, case of 6 ea bulk pack
Synonym(e):
polycarbonate Bottles, square bottle, sterile bottle
Größe auswählen
6 EACH
400,00 €
400,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
6 EACH
400,00 €
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
400,00 €
Check Cart for Availability
Materialien
clear polycarbonate
Sterilität
sterile
Verpackung
case of 6 ea bulk pack
Hersteller/Markenname
Nalgene 3405-42
Kapazität
5000 mL
5000 mL capacity
Allgemeine Beschreibung
Reduce the risk of contamination through carryover and save the cost of cleaning, sterilization and related validations by using Nalgene® Biotainer® Sterile, ready-to-use bottles and carboys designed specifically for pharmaceutical and biological manufacturers. The robustness and impact resistance of PC at cold termperatures mean these containers are ideal for freezing high-value products.
Leistungsmerkmale und Vorteile
  • Durable, translucent PC ensures sample integrity
  • White, silicone-lined, radiation stable polypropylene (PP) closures for leak resistance
  • Graduations printed on bottles and molded into containers over 5L
  • Individual customization options for fluid transfer
  • Extensive validation documents are at your disposal
  • Suitable for storage at temperatures from -100 °C to +100 °C
Rechtliche Hinweise
Biotainer is a registered trademark of In Vitro Scientific Products
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)