Merck
TMO312085-0016
Nalgene® narrow-mouth bottle
amber high-density polyethylene, size 500 mL, case of 125 ea
Synonym(e):
Nalgene bottle, plastic bottle
Größe auswählen
125 EACH
278,00 €
278,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
125 EACH
278,00 €
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
278,00 €
Check Cart for Availability
Materialien
amber high-density polyethylene
Verpackung
case of 125 ea
Hersteller/Markenname
Nalgene 312085-0016
Größe
500 mL
Allgemeine Beschreibung
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)