Merck
TMO312006-9050
Nalgene® diagnostic bottle
natural polypropylene copolymer, volume 15 mL, case of 2000 ea
Synonym(e):
PPCO bottles, small volume bottle with closure
Größe auswählen
2000 EACH
1.630,00 €
1.630,00 €
Voraussichtliches Versanddatum03. April 2025
Größe auswählen
Ansicht ändern
2000 EACH
1.630,00 €
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
1.630,00 €
Voraussichtliches Versanddatum03. April 2025
Materialien
natural polypropylene copolymer
Verpackung
case of 2000 ea
Hersteller/Markenname
Nalgene 312006-9050
Kapazität
15 mL
Volumen
15 mL
Allgemeine Beschreibung
Sample, store and ship small volume, valuable reagents. Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Bottles include linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)