Merck
TMO136196
Nunc® Cell Culture Treated Flask with filter cap
polystyrene, flask capacity 7 mL, Nunclon® Delta surface, sterile, filter cap
Synonym(e):
cell culture flask, treated flask
Größe auswählen
160 EACH
562,00 €
562,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
160 EACH
562,00 €
About This Item
UNSPSC-Code:
41161502
NACRES:
NB.13
562,00 €
Check Cart for Availability
Materialien
polystyrene
Sterilität
sterile
Verpackung
pack of 160 ea
Hersteller/Markenname
Nunc 136196
Kolbenkapazität
7 mL
Allgemeine Beschreibung
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific Nunc Cell Culture Treated Flasks with Filter Caps. These flasks receive the proprietary Nunclon Delta surface treatment to ensure consistent cell growth. The filter caps allow constant airflow and minimize chances of contamination.
Rechtliche Hinweise
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)