T1168
Nalgene® centrifuge tubes, Oak Ridge Style 3119
nominal capacity 28 mL
Synonym(e):
High-Speed Centrifuge Tubes, Nalgene tubes
Größe auswählen
110,00 €
Voraussichtliches Versanddatum20. Mai 2025
Größe auswählen
About This Item
110,00 €
Voraussichtliches Versanddatum20. Mai 2025
Empfohlene Produkte
Materialien
polypropylene copolymer
polypropylene screw closure
round bottom tube
screw top cap (Linerless)
Leistungsmerkmale
RCF 50,000 × g
Verpackung
pack of 10 ea
Hersteller/Markenname
Nalgene® 3119-0028
Nennkapazität
28 mL
Allgemeine Beschreibung
Leistungsmerkmale und Vorteile
- Exhibits higher chemical resistance compared to polycarbonate tubes.
- Can withstand repeated autoclaving, resulting in a longer lifespan compared to regular polycarbonate tubes subjected to the same conditions.
- Includes polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
- The tubes are autoclavable and become leakproof when used with the provided Nalgene closure.
Sonstige Hinweise
- Tubes should be filled to a minimum of 80% capacity to ensure proper performance.
- These centrifuge tubes are recyclable in numerous communities.
- When the product reaches the end of its life, kindly wash and recycle your Nalgene PP and PPCO centrifuge tubes if recycling services are available in your area.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)