Merck
GBL472757
Grace Bio-Labs ProPlate® microarray system, slide module only
well size 24, W × L × depth 6.8 mm × 5.4 mm × 7.6 mm, black, without clips
Synonym(e):
incubation chambers, microarray hybridization chambers, slide chambers
Größe auswählen
10 EA
265,00 €
265,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
265,00 €
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
265,00 €
Check Cart for Availability
Materialien
black silicone (on Delrin)
Sterilität
non-sterile
Leistungsmerkmale
black, without clips
Verpackung
pack of 10 ea
Hersteller/Markenname
Grace Bio-Labs 472757
B × L × Tiefe
6.8 mm × 5.4 mm × 7.6 mm
Well-Größe
24
Allgemeine Beschreibung
Grace Bio-Labs ProPlate® multi-array chambers (microarray systems) are designed to be used with ONCYTE® nitrocellulose-coated slides. These multi-array chambers integrate microscope slide based microarray technology with high-throughput (HTS applications) microtiter plate processing. Up to 64 individual arrays can be assayed per slide.
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
  • Up to 64 leak-proof wells on any slide surface without the use of adhesive
  • Microtiter plate well spacing facilitates use with multichannel pipettes
  • 7 x 7 mm square wells are ideal for high content arrays 5-350 ul well volumes accommodate small reagent and large wash volumes
Rechtliche Hinweise
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)