Merck
GBL204997
Grace Bio-Labs FlexPlate MP microtiter plate superstructure
size 96 wells, L × W 7.25 mm × 7.25 mm, clear silicone, adhesive one side, with black polystyrene frame
Synonym(e):
incubation chamber cover, silicone microtiter plate cover
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
clear silicone
Sterilität
non-sterile
Leistungsmerkmale
lid: no
Verpackung
pack of 1 ea
Hersteller/Markenname
Grace Bio-Labs 204997
L × B
7.25 mm × 7.25 mm
Größe
96 wells
Verwandte Kategorien
Allgemeine Beschreibung
The FlexPlate MP SBS compliant microtiter plate features a clean release adhesive silicone gasket which forms wells on substrates used in microplate assays. The removable silicone gasket that forms the wells of this system saves time as well depth and subsequent z-axis travel by robotics is eliminated.
  • SBS compliant microtiter plate format which enables robotic processing of assays
  • Water-tight seal between superstructure and substrate
  • Compatible with glass substrates
  • Available in 96 square well format
  • Available as a complete microtiter plate with nitrocellulose film coated substrate, uncoated substrate or as a removable superstructure alone
Rechtliche Hinweise
FlexPlate MP is a trademark of Grace Bio-Labs, Inc.