Merck
GBL104412
Grace Bio-Labs MultiSlip coverglass inserts, with chambered coverglass
L × W 12 mm × 12 mm, sterile, wells 15
Synonym(e):
chamber slides, chambered coverglass, cover slides, incubation chambers
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
Materialien
colorless
silicone (on glass)
Sterilität
sterile
Verpackung
pack of 10 ea
Hersteller/Markenname
Grace Bio-Labs 104412
L × B
12 mm × 12 mm
Wellbereich
1.44 cm2
Wells
15
Allgemeine Beschreibung
Grace Bio-Labs MultiSlip insert with 8 (18 mm x 18 mm) or 15 (12 mm x 12 mm) number 1.5 German coverglass per insert are sterile and ready to use in conventional 86 mm x 128 mm culture plates for cell culture applications. Staining and washing procedures may be performed with MultiSlip inserts in the plate, or silicone backed coverglass may be removed individually and affixed to glass microscope slides. Alternately, inserts may be easily removed for batch processing in glass staining dishes.
Note: MultiSlip coverglass inserts are ideally suited for the culture of cells where pretreatment of glass surfaces with a biological coating is required.
Rechtliche Hinweise
Multislip is a trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)