Merck
EPAG7-TT25
Test Tube Rack, for Eppendorf® New Brunswick Innova® 3100 Water Bath Shakers
holds 28 x 18 mm tubes
Größe auswählen
1 EA
405,00 €
405,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
405,00 €
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.73
405,00 €
Check Cart for Availability
Materialien
stainless steel
Leistungsmerkmale
holds 28 x 18 mm tubes
Hersteller/Markenname
Eppendorf® AG7-TT25 (North America)
Allgemeine Beschreibung
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)