Merck
EP0030129849
Eppendorf® BioBased twin.tec 96 Well PCR Plate
greener alternative
skirt, clear (with OptiTrack high contrast labelling)
About This Item
UNSPSC-Code:
41106300
NACRES:
NB.15
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
clear (with OptiTrack high contrast labelling)
polypropylene
Sterilität
non-sterile
Leistungsmerkmale
lid: no
skirt
Verpackung
pkg of 25 ea
Hersteller/Markenname
Eppendorf® 0030129849
Grünere Alternativprodukt-Eigenschaften
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
Größe
96 well
Well-Volumen
150 μL
Eignung
suitable for PCR
Grünere Alternativprodukt-Kategorie
Allgemeine Beschreibung
Eppendorf® twin.tec PCR Plate 96, BioBased, skirted, PCR clean, clear, 25 plates
Leistungsmerkmale und Vorteile
  • Reducing your environmental footprint inthe lab
  • Produced from second-generation biobased with 90% of thefeedstock attributed to renewableresources (i.e. waste and residues fromvegetable oil refining or waste cooking oil), avoiding the use of fossilfuel-based petroleum
  • Eppendorf production facility is ISCC PLUS (InternationalSustainability & Carbon Certification) certified, confirming the rawmaterials used at the facility are “bio-circular”. Enables traceability alongthe entire supply chain
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performanceExtremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contaminationBatch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)