Merck
AXYPDW500CS
Deep well plate
for SBS FootPrint, size 96 wells, size 600 μL, pkg of 10x5plates/cs, sterile
Synonym(e):
Corning cell culture plate
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien:
round bottom wells
Größe:
600 μL
96 wells
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
round bottom wells
Sterilität
sterile
Hersteller/Markenname
Axygen Inc P-DW-500-C-S
Corning
Verpackung
pkg of 10x5plates/cs
Größe
600 μL
96 wells
Allgemeine Beschreibung
  • Axygen® deep well assay plates are ideal for use in applications including sample collection, in vitro growth chambers and long-term storage.
  • Chemically resistant and withstands temperatures down to -80°C.
  • Ultra-low profile design for reduced space requirements.
  • Flat surface for efficient sealing using heat sealing film.
Rechtliche Hinweise
Axygen is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)