STC11FHA
Lynx® ST
1/4" Lynx® ST Connector, 3/4" TC inlet and 1/4" hose barb outlet
Synonym(e):
Steam-To aseptic connector. Connect hard piped equipment with single use tubing
Größe auswählen
98,00 €
Größe auswählen
About This Item
98,00 €
Empfohlene Produkte
Materialien
molded polyetherimide (21 CFR part 177.1595)
platinum cured silicone seal (21 CFR part 177.2600)
Qualitätsniveau
Sterilität
non-sterile
Produktlinie
Lynx®
Parameter
temperature (SIP : 4X at 145°C/293°F for 30 min. Closed)
1.4 bar max. pressure (20 psi) (during actuation)
2 L/min flow rate at 0.1 bar (1.5 psi)
4 L/min flow rate at 0.4 bar (6 psi)
4-80 °C operating temperature (39-176 °F)
6.89 bar max. pressure (100 psi) (operating pressure)
7 L/min flow rate at 1.3 bar (19 psi)
L × B
8.1 cm (3.2 in.) × 6.9 cm (2.7 in.)
Fitting
3/4 in. Tri-Clover inlet
1/4 in. outlet hose barb
Verwandte Kategorien
Angaben zur Herstellung
Gamma compatible up to 45 kGy; Steam @ 135 °C for 30 min (closed) 2 times
Artikel
This article explains how to streamline transfer of an allogeneic cell therapies process to a CDMO partner and long-term manufacturing success.