Merck
MIX0100L101
Millipore
Mobius® MIX100, 230 V, 50/60 Hz
About This Item
UNSPSC-Code:
41103801
eCl@ss:
32010600
Aufnahme AC/DC
230 V, 50 - 60 Hz
Qualitätsniveau
Komponenten
Includes carrier, control box unit, motor, hand-held control and cable
Rechtliche Hinweise
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
Zubehör
Produkt-Nr.
Beschreibung
Preisangaben
Die Dokumentenbibliothek aufrufen
Verwandter Inhalt
This page summarizes key considerations related to regulatory expectations for biomanufacturing including chemicals and polymeric materials. For chemicals, topics include nitrosamines and elemental impurities, and for polymeric materials such as filters and single-use assemblies, the focus is qualification and extractable and leachable testing.