Merck
Z743032
Julabo® CORIO CD heating circulator
CD-BC6, bath capacity 4.5 L (6 L), temperature 20-150 °C, stainless steel bath, AC/DC input 230 V AC, Schuko plug
Größe auswählen
1 EA
2.200,00 €
2.200,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
2.200,00 €
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
2.200,00 €
Check Cart for Availability
Materialien
stainless steel bath
Hersteller/Markenname
Julabo 9012506
Temperaturmessbereich
20-150 °C
Badkapazität
4.5 L (6 L)
Außenmaß B × L × H
24 cm × 44 cm × 47 cm
Öffnung B × L × H
13 cm × 15 cm × 20 cm
Allgemeine Beschreibung
CORIO CD heating circulators feature professional technology for high standards. These units facilitate internal temperature control in the bath tank or control of externally connected applications.
Your advantages
  • Models for internal and external applications
  • Bright, white, easy to read display
  • Very quiet
  • Easy pump change-over between internal and external circulation
  • USB connection
  • High-quality bath tanks made of stainless steel with bath lid and drain tap
  • Integrated cooling coil for counter-cooling
Rechtliche Hinweise
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)